Assay Details
| Gene Symbol: Ict1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026729.1 | NP_081005.1 | 2 - 3 | 146 | 64 | |
| GenBank mRNA | AK003192.1 | - | 2 - 3 | 146 | 64 | |
| AK003286.1 | - | 2 - 3 | 118 | 64 | ||
| BC028523.1 | - | 2 - 3 | 247 | 2938122 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ict1
Genomic Map