Assay Details
| Gene Symbol: Tm2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026795.3 | NP_081071.1 | 1 - 2 | 152 | 110 | |
| GenBank mRNA | AK003917.1 | - | 1 - 2 | 111 | 110 | |
| BC024620.1 | - | 1 - 2 | 105 | 4192712 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm2d3
Genomic Map