Assay Details
| Gene Symbol: Tm2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026795.3 | NP_081071.1 | 3 - 4 | 510 | 59 | |
| NM_178056.3 | NP_835157.1 | 4 - 5 | 603 | 59 | ||
| GenBank mRNA | AK003917.1 | - | 3 - 4 | 469 | 59 | |
| AK077858.1 | - | 1217 | 59 | |||
| BC024620.1 | - | 3 - 4 | 463 | 4192712 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm2d3
Genomic Map