| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100005411.mm.1 |
NM_001159905.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005411.mm.1 |
NM_001159905.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005411.mm.1 |
NM_001159906.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005411.mm.1 |
NM_001159906.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005411.mm.1 |
NM_026846.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005411.mm.1 |
NM_026846.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005411.mm.1 |
TC0100000653.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005408.mm.1 |
NM_001159905.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005408.mm.1 |
NM_001159905.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005408.mm.1 |
NM_001159906.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005408.mm.1 |
NM_001159906.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005408.mm.1 |
NM_026846.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005408.mm.1 |
NM_026846.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005408.mm.1 |
TC0100000653.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005407.mm.1 |
NM_001159905.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005407.mm.1 |
NM_001159905.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005407.mm.1 |
NM_001159906.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005407.mm.1 |
NM_001159906.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005407.mm.1 |
NM_026846.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005407.mm.1 |
NM_026846.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005407.mm.1 |
TC0100000653.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005414.mm.1 |
NM_001159905.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005414.mm.1 |
NM_001159905.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005414.mm.1 |
NM_001159906.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005414.mm.1 |
NM_001159906.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005414.mm.1 |
NM_026846.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005414.mm.1 |
NM_026846.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005414.mm.1 |
TC0100000653.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005413.mm.1 |
NM_001159905.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005413.mm.1 |
NM_001159905.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005413.mm.1 |
NM_001159906.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005413.mm.1 |
NM_001159906.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005413.mm.1 |
NM_026846.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005413.mm.1 |
NM_026846.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005413.mm.1 |
TC0100000653.mm.1 |
MTA TC |