Assay Details
| Gene Symbol: Zfand2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159905.1 | NP_001153377.1 | 8 - 9 | 743 | 67 | |
| NM_001159906.1 | NP_001153378.1 | 8 - 9 | 740 | 67 | ||
| NM_026846.3 | NP_081122.2 | 7 - 8 | 777 | 67 | ||
| GenBank mRNA | AK004332.1 | - | 7 - 8 | 767 | 67 | |
| AK151430.1 | - | 7 - 8 | 779 | 67 | ||
| BC011495.1 | - | 7 - 8 | 802 | 4196340 | 67 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352374 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352372 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352373 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352371 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P488718 | AK151430.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P488718 | BC011495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P488718 | BX633014.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P488718 | NM_001159905.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P488718 | NM_001159906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P488718 | AK151430.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P488718 | BC011495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P488718 | BX633014.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P488718 | NM_001159905.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P488718 | NM_001159906.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116997 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153857 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119724 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157588 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P488718 | AK151430.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P488718 | BC011495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P488718 | BX633014.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P488718 | NM_001159905.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P488718 | NM_001159906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P488718 | AK151430.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P488718 | BC011495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P488718 | BX633014.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P488718 | NM_001159905.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P488718 | NM_001159906.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352372 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157588 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153857 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119724 | - | - |
| Whole Mouse Genome G4122A | A_51_P488718 | AK151430.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P488718 | BC011495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P488718 | BX633014.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P488718 | NM_001159905.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P488718 | NM_001159906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P488718 | AK151430.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P488718 | BC011495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P488718 | BX633014.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P488718 | NM_001159905.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P488718 | NM_001159906.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123104 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113581 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157588 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119724 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153857 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00970170 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00880272 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00970162 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00880258 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00880255 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00880267 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352374 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352372 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00204407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00880261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00204385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00880266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00904000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00970167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00880271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00204398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00204392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00880267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352387 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10347524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347527 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347528 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347525 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451390_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424521_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10347521 | - | - |
| Mouse Genome 430 2.0 Array | 1451390_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1424521_at | - | - |
| Mouse Genome 430 2.0 Array | 1424521_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451390_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68818_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68818_PM_at | - | - |
| Mouse Expression Set 430 | 1424521_at | - | - |
| Mouse Expression Set 430 | 1451390_s_at | - | - |
| Mouse Expression Set 430 | 1451390_PM_s_at | - | - |
| Mouse Expression Set 430 | 1424521_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451390_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424521_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451390_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424521_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424521_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424521_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451390_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451390_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 650722 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2480619 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6650047 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5050156 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2480619 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5050156 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6650047 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005050156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002480619 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006650047 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2763951 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2659440 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2676634 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2676634 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2659440 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2763951 | - | - |
| mouse-6 v1.1 (Illumina) | scl17782.7.1_158-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000808.1_59-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000926.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5050156 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2480619 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6650047 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000926.1_1-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000808.1_59-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17782.7.1_158-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480619 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5050156 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650047 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2676634 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2659440 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2800675 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2763951 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2676634 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2800675 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2763951 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2659440 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000926.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000808.1_59-S | - | - |

Zfand2b
Genomic Map