Assay Details
| Gene Symbol: Xrcc6bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159559.1 | NP_001153031.1 | 5 - 6 | 619 | 76 | |
| GenBank mRNA | AK004405.1 | - | 5 - 6 | 599 | 76 | |
| CB203044.1 | - | 2 - 3 | 234 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Xrcc6bp1
Genomic Map