Assay Details
| Gene Symbol: Krt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027011.2 | NP_081287.1 | 7 - 8 | 1498 | 106 | |
| GenBank mRNA | AK014363.1 | - | 2 - 3 | 282 | 106 | |
| BC006780.1 | - | 7 - 8 | 1498 | 3589013 | 106 | |
| BC108361.1 | - | 7 - 8 | 1495 | 6312495 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt5
Genomic Map