Assay Details
| Gene Symbol: Avpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027106.4 | NP_081382.1 | 1 - 2 | 117 | 104 | |
| GenBank mRNA | AK009243.1 | - | 1 - 2 | 118 | 104 | |
| BC027646.1 | - | 1 - 2 | 151 | 1344413 | 104 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK009243.1,BC027646.1,NM_027106.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119206 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119205 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119202 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119207 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119203 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119204 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120254 | NM_027106.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120254 | AK009243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120254 | AK177091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120254 | AY505175.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120254 | BC027646.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692187 | AK003884.3 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692187 | AK048669.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692187 | AK154192.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692187 | AK164964.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692187 | BC042570.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692187 | NM_176833.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462146 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100974 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180389 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120254 | NM_027106.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120254 | AK009243.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120254 | AK177091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120254 | AY505175.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120254 | BC027646.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P261087 | NM_027106.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P261087 | AK009243.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P261087 | AY505175.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119232 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180389 | - | - |
| Whole Mouse Genome G4122A | A_51_P261087 | NM_027106.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P261087 | AK009243.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P261087 | AY505175.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132362 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180389 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14245449 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14245462 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14245454 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14245455 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14245446 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14245463 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119205 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119204 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119206 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119202 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119207 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32119191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14225309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14245449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14151185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14363697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14475321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14151186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14245445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14219441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14151176 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10467743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467740 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423122_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10467739 | - | - |
| Mouse Genome 430 2.0 Array | 1423122_at | - | - |
| Mouse Genome 430 2.0 Array | 1423122_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69534_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69534_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900013427.mm.1 | TC1900001521.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900013425.mm.1 | NM_027106.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900013425.mm.1 | NM_027106.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900013425.mm.1 | TC1900001521.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423122_PM_at | - | - |
| Mouse Expression Set 430 | 1423122_at | - | - |
| Murine Genome U74v2 Set | 96866_at | - | - |
| Murine Genome U74v2 Set | 96866_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423122_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423122_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423122_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423122_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 523824 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1850377 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001850377 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2755585 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2755585 | - | - |
| mouse-6 v1.1 (Illumina) | scl52477.5.1_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1850377 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52477.5.1_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850377 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2813186 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2755585 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2813186 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2755585 | - | - |

Avpi1
Genomic Map