Assay Details
| Gene Symbol: Rnf215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027859.2 | NP_082135.2 | 5 - 6 | 862 | 86 | |
| GenBank mRNA | AK002414.1 | - | 5 - 6 | 820 | 86 | |
| AK047194.1 | - | 811 | 86 | |||
| AK182091.1 | - | 4 - 5 | 244 | 86 | ||
| BC103627.1 | - | 5 - 6 | 786 | 6486672 | 86 | |
| BC115907.1 | - | 5 - 6 | 809 | 40105152 | 86 | |
| BC115908.1 | - | 5 - 6 | 809 | 40105148 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555852 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555851 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166262 | BC103627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166262 | BM120893.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166262 | BM948157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166262 | AK002414.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166262 | AK154140.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519420 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649802 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 948628 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 107485 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103287 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166262 | BC103627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166262 | BM120893.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166262 | BM948157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166262 | AK002414.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166262 | AK154140.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555864 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07239919 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103287 | - | - |
| Whole Mouse Genome G4122A | A_51_P208019 | BC103627.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208019 | BM120893.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208019 | BM948157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208019 | AK002414.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208019 | AK154140.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103268 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103287 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09466793 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09466786 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09466787 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09466791 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09352930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09352935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09466791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09589188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09105124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09352940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09465126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09105110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09352936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09352934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27555832 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373870 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373864 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373867 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373865 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373866 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373863 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373868 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373869 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373862 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373871 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452756_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373861 | - | - |
| Mouse Genome 430 2.0 Array | 1452756_at | - | - |
| Mouse Genome 430 2.0 Array | 1452756_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71673_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71673_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100000437.mm.1 | NM_027859.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000437.mm.1 | NM_027859.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000437.mm.1 | TC1100000035.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100000436.mm.1 | NM_027859.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000436.mm.1 | NM_027859.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000436.mm.1 | TC1100000035.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1452756_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452756_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 784349 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4540575 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4540575 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004540575 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2718462 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2718462 | - | - |
| mouse-6 v1.1 (Illumina) | scl41892.7.15_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4540575 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41892.7.15_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540575 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2718462 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2718462 | - | - |

Rnf215
Genomic Map