Assay Details
| Gene Symbol: Telo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163661.1 | NP_001157133.1 | 8 - 9 | 1711 | 53 | |
| NM_027880.2 | NP_082156.2 | 8 - 9 | 1622 | 53 | ||
| GenBank mRNA | AK004582.1 | - | 8 - 9 | 1492 | 53 | |
| AK028658.1 | - | 8 - 9 | 1476 | 53 | ||
| AK045321.1 | - | 1 - 2 | 1158 | 53 | ||
| AK050773.1 | - | 8 - 9 | 1441 | 53 | ||
| AK084830.1 | - | 8 - 9 | 1492 | 53 | ||
| AK143827.1 | - | 8 - 9 | 1712 | 53 | ||
| AK146963.1 | - | 8 - 9 | 1498 | 53 | ||
| AK154727.1 | - | 8 - 9 | 1524 | 53 | ||
| AK158181.1 | - | 13 - 14 | 53 | |||
| BC011077.1 | - | 8 - 9 | 1472 | 3984911 | 53 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK045321.1 transcripts.
Set Membership: |
|

Telo2
Genomic Map