Assay Details
| Gene Symbol: Ivns1abp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039511.1 | NP_001034600.1 | 3 - 4 | 615 | 61 | |
| NM_001039512.1 | NP_001034601.1 | 3 - 4 | 615 | 61 | ||
| NM_054102.2 | NP_473443.2 | 3 - 4 | 615 | 61 | ||
| GenBank mRNA | AB055737.1 | - | 3 - 4 | 611 | 61 | |
| AB055738.1 | - | 3 - 4 | 611 | 61 | ||
| AK007291.1 | - | 3 - 4 | 565 | 61 | ||
| AK129229.1 | - | 3 - 4 | 553 | 61 | ||
| AK144619.1 | - | 3 - 4 | 594 | 61 | ||
| AK144835.1 | - | 3 - 4 | 596 | 61 | ||
| AK145071.1 | - | 3 - 4 | 606 | 61 | ||
| AK145845.1 | - | 3 - 4 | 566 | 61 | ||
| AK146001.1 | - | 3 - 4 | 596 | 61 | ||
| AK146494.1 | - | 3 - 4 | 568 | 61 | ||
| AK146645.1 | - | 3 - 4 | 566 | 61 | ||
| AK159254.1 | - | 3 - 4 | 597 | 61 | ||
| AK179875.1 | - | 2 - 3 | 166 | 61 | ||
| AK181493.1 | - | 2 - 3 | 192 | 61 | ||
| BC004092.1 | - | 2 - 3 | 315 | 3593827 | 61 | |
| BC040250.1 | - | 3 - 4 | 607 | 61 | ||
| DQ914522.1 | - | 3 - 4 | 567 | 61 | ||
| DQ914523.1 | - | 3 - 4 | 537 | 61 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the DQ914523.1 transcripts.
Set Membership: |
|

Ivns1abp
Genomic Map