Assay Details
| Gene Symbol: Eif3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285942.1 | NP_001272871.1 | 4 - 5 | 473 | 78 | |
| NM_001285943.1 | NP_001272872.1 | 4 - 5 | 473 | 78 | ||
| NM_028659.3 | NP_082935.1 | 5 - 6 | 572 | 78 | ||
| GenBank mRNA | AK004664.1 | - | 5 - 6 | 499 | 78 | |
| AK037834.1 | - | 5 - 6 | 468 | 78 | ||
| AK158875.1 | - | 4 - 5 | 450 | 78 | ||
| BC027638.1 | - | 4 - 5 | 372 | 1348838 | 78 | |
| BC091749.1 | - | 5 - 6 | 476 | 3661337 | 78 | |
| CD802664.1 | - | 3 - 4 | 78 | |||
More Information
Set Membership: |
|

Eif3k
Genomic Map