Assay Details
| Gene Symbol: Ift57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028680.3 | NP_082956.2 | 2 - 3 | 420 | 109 | |
| GenBank mRNA | AF274590.1 | - | 2 - 3 | 438 | 109 | |
| AK014731.1 | - | 2 - 3 | 472 | 109 | ||
| AK047217.1 | - | 2 - 3 | 420 | 109 | ||
| BC138143.1 | - | 2 - 3 | 394 | 8861162 | 109 | |
| BC138144.1 | - | 2 - 3 | 394 | 8861163 | 109 | |
| DQ266091.1 | - | 2 - 3 | 461 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ift57
Genomic Map