Assay Details
| Gene Symbol: Grip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277292.1 | NP_001264221.1 | 12 - 13 | 1864 | 90 | |
| NM_001277293.1 | NP_001264222.1 | 13 - 14 | 2020 | 90 | ||
| NM_001277294.1 | NP_001264223.1 | 12 - 13 | 1721 | 90 | ||
| NM_001277295.1 | NP_001264224.1 | 3 - 4 | 656 | 90 | ||
| NM_028736.2 | NP_083012.1 | 13 - 14 | 1877 | 90 | ||
| NM_130891.2 | NP_570961.1 | 11 - 12 | 1640 | 90 | ||
| NM_133442.2 | NP_597699.1 | 11 - 12 | 1783 | 90 | ||
| NR_102387.1 | 3 - 4 | 656 | 90 | |||
| GenBank mRNA | AB051560.1 | - | 11 - 12 | 1517 | 90 | |
| AB051561.1 | - | 13 - 14 | 1754 | 90 | ||
| AB051562.1 | - | 11 - 12 | 1503 | 90 | ||
| AK016420.1 | - | 3 - 4 | 654 | 90 | ||
| AK029845.1 | - | 12 - 13 | 2377 | 90 | ||
| AK029905.1 | - | 3 - 4 | 657 | 90 | ||
| AK039673.1 | - | 11 - 12 | 1525 | 90 | ||
| AK220553.1 | - | 12 - 13 | 1578 | 90 | ||
| AY255674.1 | - | 3 - 4 | 472 | 90 | ||
| AY294283.1 | - | 3 - 4 | 453 | 90 | ||
| BC067398.1 | - | 12 - 13 | 1721 | 30096759 | 90 | |
| BC072632.1 | - | 13 - 14 | 2020 | 6843802 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grip1
Genomic Map