Assay Details
| Gene Symbol: Xpo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028816.2 | NP_083092.2 | 14 - 15 | 2368 | 57 | |
| GenBank mRNA | AB093235.1 | - | 9 - 10 | 2633 | 57 | |
| AY029528.1 | - | 13 - 14 | 1864 | 57 | ||
| BC058090.1 | - | 14 - 15 | 2368 | 6826356 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xpo6
Genomic Map