Assay Details
| Gene Symbol: Xpo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028816.2 | NP_083092.2 | 17 - 18 | 2845 | 101 | |
| GenBank mRNA | AB093235.1 | - | 12 - 13 | 3110 | 101 | |
| AK154709.1 | - | 17 - 18 | 2890 | 101 | ||
| AK181559.1 | - | 1 - 2 | 64 | 101 | ||
| AK184691.1 | - | 1 - 2 | 64 | 101 | ||
| AY029528.1 | - | 16 - 17 | 2341 | 101 | ||
| BC058090.1 | - | 17 - 18 | 2845 | 6826356 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xpo6
Genomic Map