Assay Details
| Gene Symbol: Xpo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028816.2 | NP_083092.2 | 19 - 20 | 3118 | 78 | |
| GenBank mRNA | AB093235.1 | - | 14 - 15 | 3383 | 78 | |
| AK011324.2 | - | 2 - 3 | 255 | 78 | ||
| AK154709.1 | - | 19 - 20 | 3163 | 78 | ||
| AK181559.1 | - | 3 - 4 | 337 | 78 | ||
| AK184691.1 | - | 3 - 4 | 338 | 78 | ||
| AY029528.1 | - | 18 - 19 | 2614 | 78 | ||
| BC058090.1 | - | 19 - 20 | 3118 | 6826356 | 78 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK011324.2 transcripts.
Set Membership: |
|

Xpo6
Genomic Map