Assay Details
| Gene Symbol: Rita1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029096.3 | NP_083372.1 | 2 - 3 | 668 | 75 | |
| NM_133908.1 | NP_598669.1 | 3 - 4 | 763 | 75 | ||
| GenBank mRNA | AK003581.1 | - | 2 - 3 | 649 | 75 | |
| BC021365.1 | - | 2 - 3 | 567 | 5030737 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24039805 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24039806 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24039807 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24039808 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510722 | NM_133908.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510722 | NM_029096.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510722 | AK003581.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510722 | BC021365.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510722 | NM_133908.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510722 | NM_029096.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510722 | AK003581.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510722 | BC021365.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794699 | AK008126.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794699 | AK008185.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794699 | AK008395.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260551 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149255 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120901 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510722 | NM_133908.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510722 | NM_029096.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510722 | AK003581.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510722 | BC021365.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510722 | NM_133908.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510722 | NM_029096.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510722 | AK003581.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510722 | BC021365.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24039792 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149255 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120901 | - | - |
| Whole Mouse Genome G4122A | A_51_P510722 | NM_133908.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P510722 | NM_029096.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P510722 | AK003581.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P510722 | BC021365.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P510722 | NM_133908.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P510722 | NM_029096.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P510722 | AK003581.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P510722 | BC021365.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131795 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120901 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149255 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04333196 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04333193 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04333191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04333200 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24039806 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24039807 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24039805 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24039808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04487382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04216805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04371724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04446476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04487379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24039779 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10533177 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533179 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419333_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10533176 | - | - |
| Mouse Genome 430 2.0 Array | 1419333_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419333_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100764_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100764_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500028130.mm.1 | TC0500003171.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028130.mm.1 | NM_029096.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028130.mm.1 | NM_029096.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028130.mm.1 | NM_133908.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028130.mm.1 | NM_133908.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028131.mm.1 | TC0500003171.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028131.mm.1 | NM_029096.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028131.mm.1 | NM_029096.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028131.mm.1 | NM_133908.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028131.mm.1 | NM_133908.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028134.mm.1 | TC0500003171.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028134.mm.1 | NM_133908.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028134.mm.1 | NM_133908.1 | Refseq |
| Mouse Expression Set 430 | 1419333_PM_at | - | - |
| Mouse Expression Set 430 | 1419333_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419333_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419333_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419333_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419333_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 558461 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2120164 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2120164 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370593 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370593 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002120164 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2642876 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2646001 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2642876 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2646001 | - | - |
| mouse-6 v1.1 (Illumina) | scl26113.1.36_35-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0100764.1_73-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370593 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2120164 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0100764.1_73-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26113.1.36_35-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120164 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370593 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2642876 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2803878 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2646001 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2646001 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2803878 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2642876 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl26113.1.36_35-S | - | - |

Rita1
Genomic Map