Assay Details
| Gene Symbol: Lmbr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029098.3 | NP_083374.1 | 4 - 5 | 509 | 62 | |
| GenBank mRNA | AK003656.1 | - | 4 - 5 | 441 | 62 | |
| AK030589.1 | - | 4 - 5 | 475 | 62 | ||
| AK049110.1 | - | 4 - 5 | 510 | 62 | ||
| AK143891.1 | - | 4 - 5 | 470 | 62 | ||
| AK154968.1 | - | 4 - 5 | 643 | 62 | ||
| AK170328.1 | - | 4 - 5 | 470 | 62 | ||
| AK173121.1 | - | 4 - 5 | 890 | 62 | ||
| AY052398.1 | - | 4 - 5 | 352 | 62 | ||
| BC023397.1 | - | 4 - 5 | 446 | 5043877 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmbr1l
Genomic Map