Assay Details
| Gene Symbol: Lmbr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029098.3 | NP_083374.1 | 10 - 11 | 1034 | 108 | |
| GenBank mRNA | AK003656.1 | - | 10 - 11 | 966 | 108 | |
| AK030589.1 | - | 10 - 11 | 1000 | 108 | ||
| AK049110.1 | - | 10 - 11 | 1035 | 108 | ||
| AK143891.1 | - | 10 - 11 | 995 | 108 | ||
| AK154968.1 | - | 10 - 11 | 1168 | 108 | ||
| AK173121.1 | - | 10 - 11 | 1415 | 108 | ||
| AY052398.1 | - | 10 - 11 | 877 | 108 | ||
| BC023397.1 | - | 10 - 11 | 971 | 5043877 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmbr1l
Genomic Map