Assay Details
| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029291.1 | NP_083567.1 | 10 - 11 | 1062 | 118 | |
| GenBank mRNA | AK051387.1 | - | 10 - 11 | 1111 | 118 | |
| AK148105.1 | - | 10 - 11 | 1094 | 118 | ||
| AK180981.1 | - | 2 - 3 | 153 | 118 | ||
| BC013537.1 | - | 10 - 11 | 1062 | 4240784 | 118 | |
| BC024840.1 | - | 3 - 4 | 213 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ascc2
Genomic Map