Assay Details
| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029291.1 | NP_083567.1 | 11 - 12 | 1136 | 127 | |
| GenBank mRNA | AK051387.1 | - | 11 - 12 | 1185 | 127 | |
| AK148105.1 | - | 11 - 12 | 1168 | 127 | ||
| AK180981.1 | - | 3 - 4 | 227 | 127 | ||
| BC013537.1 | - | 11 - 12 | 1136 | 4240784 | 127 | |
| BC024840.1 | - | 4 - 5 | 287 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ascc2
Genomic Map