Assay Details
| Gene Symbol: Med25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029365.2 | NP_083641.1 | 9 - 10 | 1103 | 124 | |
| GenBank mRNA | AK089940.1 | - | 9 - 10 | 1107 | 124 | |
| AK090026.1 | - | 9 - 10 | 1130 | 125 | ||
| AK149867.1 | - | 10 - 11 | 1375 | 124 | ||
| AK165760.1 | - | 9 - 10 | 1152 | 124 | ||
| BC021333.1 | - | 9 - 10 | 1138 | 5027401 | 124 | |
| BC031138.1 | - | 9 - 10 | 1146 | 4972378 | 124 | |
| BC071206.1 | - | 8 - 9 | 1180 | 4925452 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med25
Genomic Map