Assay Details
| Gene Symbol: Med25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029365.2 | NP_083641.1 | 10 - 11 | 1235 | 89 | |
| GenBank mRNA | AK011662.1 | - | 1 - 2 | 134 | 89 | |
| AK089940.1 | - | 10 - 11 | 1239 | 89 | ||
| AK090026.1 | - | 10 - 11 | 1262 | 89 | ||
| AK149867.1 | - | 11 - 12 | 1507 | 89 | ||
| AK158238.1 | - | 10 - 11 | 1301 | 89 | ||
| AK165760.1 | - | 10 - 11 | 1284 | 89 | ||
| BC021333.1 | - | 10 - 11 | 1270 | 5027401 | 89 | |
| BC031138.1 | - | 10 - 11 | 1278 | 4972378 | 89 | |
| BC071206.1 | - | 9 - 10 | 1312 | 4925452 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med25
Genomic Map