Assay Details
| Gene Symbol: Unc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029770.2 | NP_084046.2 | 3 - 4 | 865 | 58 | |
| GenBank mRNA | AJ487853.1 | - | 3 - 4 | 768 | 58 | |
| AK018177.1 | - | 3 - 4 | 865 | 58 | ||
| AK147603.1 | - | 3 - 4 | 760 | 58 | ||
| AK154271.1 | - | 3 - 4 | 865 | 58 | ||
| AK154287.1 | - | 3 - 4 | 868 | 58 | ||
| AK158284.1 | - | 3 - 4 | 865 | 58 | ||
| AK170513.1 | - | 3 - 4 | 864 | 58 | ||
| BC048162.1 | - | 3 - 4 | 672 | 58 | ||
| BC057560.1 | - | 3 - 4 | 672 | 6417563 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc5b
Genomic Map