Assay Details
| Gene Symbol: Unc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029770.2 | NP_084046.2 | 10 - 11 | 2099 | 124 | |
| GenBank mRNA | AJ487853.1 | - | 10 - 11 | 2002 | 124 | |
| AK018177.1 | - | 10 - 11 | 2099 | 124 | ||
| AK154271.1 | - | 10 - 11 | 2099 | 124 | ||
| AK154287.1 | - | 10 - 11 | 2102 | 124 | ||
| AK158284.1 | - | 10 - 11 | 2099 | 124 | ||
| BC048162.1 | - | 9 - 10 | 1873 | 124 | ||
| BC057560.1 | - | 9 - 10 | 1873 | 6417563 | 124 | |
More Information
Set Membership: |
|

Unc5b
Genomic Map