Assay Details
| Gene Symbol: Rnft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029788.5 | NP_084064.1 | 6 - 7 | 1071 | 84 | |
| GenBank mRNA | AK002624.1 | - | 6 - 7 | 1029 | 84 | |
| AK036401.1 | - | 6 - 7 | 1072 | 84 | ||
| AK153293.1 | - | 6 - 7 | 1040 | 84 | ||
| AK171077.1 | - | 5 - 6 | 2322 | 84 | ||
| BC037057.1 | - | 5 - 6 | 858 | 84 | ||
| BC050796.1 | - | 6 - 7 | 1039 | 6743413 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnft1
Genomic Map