Assay Details
| Gene Symbol: Cryl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030004.3 | NP_084280.2 | 6 - 7 | 775 | 74 | |
| GenBank mRNA | AB359906.1 | - | 6 - 7 | 740 | 74 | |
| AF351609.1 | - | 6 - 7 | 821 | 74 | ||
| AK043569.1 | - | 6 - 7 | 822 | 74 | ||
| AK080625.1 | - | 6 - 7 | 796 | 74 | ||
| AK135834.1 | - | 6 - 7 | 789 | 74 | ||
| BC004074.1 | - | 6 - 7 | 775 | 3591359 | 74 | |
| BC027064.1 | - | 6 - 7 | 792 | 5355916 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cryl1
Genomic Map