Assay Details
| Gene Symbol: Wrnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030215.3 | NP_084491.3 | 1 - 2 | 1017 | 95 | |
| GenBank mRNA | AB056151.1 | - | 1 - 2 | 866 | 95 | |
| AF208046.1 | - | 1 - 2 | 262 | 95 | ||
| AK041886.1 | - | 1 - 2 | 973 | 95 | ||
| AK050368.1 | - | 1 - 2 | 1019 | 95 | ||
| AK082078.1 | - | 1 - 2 | 932 | 95 | ||
| AK167570.1 | - | 1 - 2 | 1018 | 95 | ||
| AK170542.1 | - | 1 - 2 | 1010 | 95 | ||
| AK170846.1 | - | 1 - 2 | 998 | 95 | ||
| BC010482.1 | - | 1 - 2 | 1009 | 3492125 | 95 | |
| BC058744.1 | - | 1 - 2 | 407 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wrnip1
Genomic Map