Assay Details
| Gene Symbol: Dcaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165256.1 | NP_001158728.1 | 2 - 3 | 292 | 88 | |
| NM_030246.2 | NP_084522.1 | 3 - 4 | 349 | 88 | ||
| GenBank mRNA | BC003284.1 | - | 2 - 3 | 258 | 3582095 | 88 |
| CN722277.1 | - | 2 - 3 | 267 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf4
Genomic Map