Assay Details
| Gene Symbol: Dcaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165256.1 | NP_001158728.1 | 9 - 10 | 1005 | 87 | |
| NM_030246.2 | NP_084522.1 | 10 - 11 | 1062 | 87 | ||
| GenBank mRNA | BC003284.1 | - | 9 - 10 | 971 | 3582095 | 87 |
| CX208460.1 | - | 4 - 5 | 277 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf4
Genomic Map