Assay Details
| Gene Symbol: Dcaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165256.1 | NP_001158728.1 | 10 - 11 | 1111 | 84 | |
| NM_030246.2 | NP_084522.1 | 11 - 12 | 1168 | 84 | ||
| GenBank mRNA | BC003284.1 | - | 10 - 11 | 1077 | 3582095 | 84 |
| CX208460.1 | - | 5 - 6 | 383 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf4
Genomic Map