Assay Details
| Gene Symbol: Dcaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165256.1 | NP_001158728.1 | 11 - 12 | 1288 | 116 | |
| NM_030246.2 | NP_084522.1 | 12 - 13 | 1345 | 116 | ||
| GenBank mRNA | BC003284.1 | - | 11 - 12 | 1254 | 3582095 | 116 |
| CX208460.1 | - | 6 - 7 | 560 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf4
Genomic Map