Assay Details
| Gene Symbol: Kank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030697.2 | NP_109622.1 | 2 - 3 | 99 | 84 | |
| GenBank mRNA | AK002623.1 | - | 2 - 3 | 100 | 84 | |
| KJ650203.1 | - | 1 - 2 | 30 | 84 | ||
| KJ663714.1 | - | 1 - 2 | 30 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kank3
Genomic Map