Assay Details
| Gene Symbol: Myb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198914.1 | NP_001185843.1 | 7 - 8 | 1107 | 95 | |
| NM_010848.3 | NP_034978.3 | 7 - 8 | 1107 | 95 | ||
| GenBank mRNA | AK088079.1 | - | 7 - 8 | 1028 | 95 | |
| AK088598.1 | - | 7 - 8 | 1029 | 95 | ||
| AK132250.1 | - | 7 - 8 | 972 | 95 | ||
| AK137965.1 | - | 7 - 8 | 973 | 95 | ||
| BC011513.1 | - | 7 - 8 | 1015 | 3672769 | 95 | |
| X02774.1 | - | 7 - 8 | 877 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myb
Genomic Map