Assay Details
| Gene Symbol: Atp6v0b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033617.3 | NP_291095.1 | 2 - 3 | 209 | 72 | |
| GenBank mRNA | AB060654.1 | - | 2 - 3 | 170 | 72 | |
| AF356006.1 | - | 2 - 3 | 213 | 72 | ||
| AK009495.1 | - | 2 - 3 | 201 | 72 | ||
| AK021272.1 | - | 2 - 3 | 189 | 72 | ||
| AK151365.1 | - | 2 - 3 | 181 | 72 | ||
| AK151847.1 | - | 2 - 3 | 197 | 72 | ||
| AK152330.1 | - | 2 - 3 | 214 | 72 | ||
| AK152823.1 | - | 2 - 3 | 197 | 72 | ||
| AK200953.1 | - | 2 - 3 | 123 | 72 | ||
| AK217120.1 | - | 2 - 3 | 165 | 72 | ||
| BC009169.1 | - | 2 - 3 | 210 | 2812497 | 72 | |
| BC030393.1 | - | 2 - 3 | 194 | 4922226 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0b
Genomic Map