Assay Details
| Gene Symbol: Atp6v0b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033617.3 | NP_291095.1 | 4 - 5 | 363 | 135 | |
| GenBank mRNA | AB060654.1 | - | 4 - 5 | 324 | 135 | |
| AF356006.1 | - | 4 - 5 | 367 | 135 | ||
| AK009495.1 | - | 4 - 5 | 354 | 134 | ||
| AK021272.1 | - | 4 - 5 | 343 | 135 | ||
| AK151365.1 | - | 4 - 5 | 334 | 135 | ||
| AK151847.1 | - | 4 - 5 | 351 | 135 | ||
| AK152330.1 | - | 4 - 5 | 368 | 135 | ||
| AK152823.1 | - | 4 - 5 | 351 | 135 | ||
| BC030393.1 | - | 4 - 5 | 348 | 4922226 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0b
Genomic Map