Assay Details
| Gene Symbol: Mrpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039084.1 | NP_001034173.1 | 4 - 5 | 551 | 97 | |
| NM_053158.3 | NP_444388.2 | 4 - 5 | 551 | 97 | ||
| GenBank mRNA | AB049632.1 | - | 3 - 4 | 507 | 97 | |
| AK015476.1 | - | 4 - 5 | 511 | 97 | ||
| AK017938.1 | - | 4 - 5 | 523 | 97 | ||
| AK083978.1 | - | 4 - 5 | 527 | 97 | ||
| AK142249.1 | - | 4 - 5 | 526 | 97 | ||
| AK172544.1 | - | 4 - 5 | 516 | 97 | ||
| BC061042.1 | - | 4 - 5 | 515 | 6706599 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl1
Genomic Map