Assay Details
| Gene Symbol: Sp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF184902.1 | - | 1 - 2 | 115 | 137 | |
| AK032521.1 | - | 1 - 2 | 108 | 137 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF184902.1, AK032521.1 transcripts.
Set Membership: |
|

Sp7
Genomic Map