Assay Details
| Gene Symbol: Adarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024837.2 | NP_001020008.1 | 3 - 4 | 424 | 101 | |
| NM_130895.3 | NP_570965.2 | 3 - 4 | 424 | 101 | ||
| GenBank mRNA | AF291049.1 | - | 2 - 3 | 259 | 101 | |
| AF403106.2 | - | 3 - 4 | 376 | 101 | ||
| AF403107.2 | - | 3 - 4 | 376 | 101 | ||
| AK138318.1 | - | 3 - 4 | 417 | 101 | ||
| AK141777.1 | - | 3 - 4 | 396 | 101 | ||
| AK147298.1 | - | 4 - 5 | 503 | 101 | ||
| FJ169505.1 | - | 2 - 3 | 199 | 101 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF291049.1,AF403106.2,AF403107.2,AK138318.1,AK141777.1,AK147298.1,NM_001024837.2,NM_130895.3 transcripts.
Set Membership: |
|

Adarb1
Genomic Map