Assay Details
| Gene Symbol: Adarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024837.2 | NP_001020008.1 | 5 - 6 | 1478 | 68 | |
| NM_130895.3 | NP_570965.2 | 5 - 6 | 1478 | 68 | ||
| NR_004429.1 | 5 - 6 | 1525 | 68 | |||
| NR_021486.1 | 5 - 6 | 1525 | 68 | |||
| GenBank mRNA | AF291049.1 | - | 4 - 5 | 1313 | 68 | |
| AF403106.2 | - | 5 - 6 | 1430 | 68 | ||
| AF403107.2 | - | 5 - 6 | 1430 | 68 | ||
| AF403108.2 | - | 5 - 6 | 1477 | 68 | ||
| AF403109.2 | - | 5 - 6 | 1477 | 68 | ||
| AF525421.1 | - | 6 - 7 | 1503 | 68 | ||
| AK141777.1 | - | 5 - 6 | 1450 | 68 | ||
| AK147298.1 | - | 6 - 7 | 1557 | 68 | ||
| AY162454.1 | - | 6 - 7 | 1503 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adarb1
Genomic Map