Assay Details
| Gene Symbol: Adarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024837.2 | NP_001020008.1 | 6 - 7 | 1633 | 89 | |
| NM_130895.3 | NP_570965.2 | 6 - 7 | 1633 | 89 | ||
| NR_004429.1 | 6 - 7 | 1680 | 89 | |||
| NR_021486.1 | 6 - 7 | 1680 | 89 | |||
| GenBank mRNA | AF291049.1 | - | 5 - 6 | 1468 | 89 | |
| AF403106.2 | - | 6 - 7 | 1585 | 89 | ||
| AF403107.2 | - | 6 - 7 | 1585 | 89 | ||
| AF403108.2 | - | 6 - 7 | 1632 | 89 | ||
| AF403109.2 | - | 6 - 7 | 1632 | 89 | ||
| AF525421.1 | - | 7 - 8 | 1658 | 89 | ||
| AK141777.1 | - | 6 - 7 | 1605 | 89 | ||
| AK147298.1 | - | 7 - 8 | 1712 | 89 | ||
| AY162454.1 | - | 7 - 8 | 1658 | 89 | ||
| BC040200.1 | - | 2 - 3 | 188 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adarb1
Genomic Map