Assay Details
| Gene Symbol: Arr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133205.3 | NP_573468.1 | 4 - 5 | 208 | 88 | |
| GenBank mRNA | AF156979.1 | - | 4 - 5 | 149 | 88 | |
| BC017145.1 | - | 4 - 5 | 143 | 4506006 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arr3
Genomic Map