Assay Details
| Gene Symbol: Rbm39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133242.3 | NP_573505.2 | 12 - 13 | 1542 | 68 | |
| GenBank mRNA | AK147076.1 | - | 12 - 13 | 1487 | 68 | |
| BC004000.1 | - | 8 - 9 | 874 | 68 | ||
| BC023205.1 | - | 12 - 13 | 1444 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbm39
Genomic Map