Assay Details
| Gene Symbol: Ppargc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133249.2 | NP_573512.1 | 1 - 2 | 140 | 64 | |
| GenBank mRNA | AF453324.1 | - | 1 - 2 | 140 | 64 | |
| AK086111.1 | - | 1 - 2 | 135 | 64 | ||
| BC150699.1 | - | 1 - 2 | 120 | 9087610 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
The orphan nuclear receptor, RORalpha, regulates gene expression that controls lipid metabolism: staggerer (SG/SG) mice are resistant to diet-induced obesity.
- Authors
- Lau P, Fitzsimmons RL, Raichur S, Wang SC, Lechtken A, Muscat GE
- Journal
- J Biol Chem - Page No 18411 (Fri Jun 27 00:00:00 EDT 2008)
- Volume (Year)
- 283 (2008)
- Product Usage
- -

Ppargc1b
Genomic Map