Assay Details
| Gene Symbol: Ppargc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133249.2 | NP_573512.1 | 3 - 4 | 534 | 75 | |
| GenBank mRNA | AF453324.1 | - | 3 - 4 | 534 | 75 | |
| AK028464.1 | - | 4 - 5 | 584 | 75 | ||
| AK086111.1 | - | 3 - 4 | 529 | 75 | ||
| BC150699.1 | - | 3 - 4 | 514 | 9087610 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppargc1b
Genomic Map