| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500031989.mm.1 |
TC0500003495.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500031989.mm.1 |
NM_001159908.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031989.mm.1 |
NM_001159908.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031989.mm.1 |
NM_133349.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031989.mm.1 |
NM_133349.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031988.mm.1 |
TC0500003495.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500031988.mm.1 |
NM_001159908.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031988.mm.1 |
NM_001159908.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031988.mm.1 |
NM_133349.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031988.mm.1 |
NM_133349.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031986.mm.1 |
TC0500003495.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500031986.mm.1 |
NM_001159908.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031986.mm.1 |
NM_001159908.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031986.mm.1 |
NM_133349.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500031986.mm.1 |
NM_133349.3 |
Refseq |