Assay Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159908.1 | NP_001153380.1 | 2 - 3 | 309 | 63 | |
| NM_133349.3 | NP_579927.1 | 2 - 3 | 320 | 63 | ||
| GenBank mRNA | AF224494.1 | - | 2 - 3 | 206 | 63 | |
| AK146082.1 | - | 2 - 3 | 258 | 63 | ||
| AK171245.1 | - | 2 - 3 | 260 | 63 | ||
| BC058780.1 | - | 2 - 3 | 246 | 6312202 | 63 | |
| CJ148035.1 | - | 2 - 3 | 315 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfand2a
Genomic Map