Assay Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159908.1 | NP_001153380.1 | 3 - 4 | 407 | 63 | |
| NM_133349.3 | NP_579927.1 | 3 - 4 | 418 | 63 | ||
| GenBank mRNA | AF224494.1 | - | 3 - 4 | 304 | 63 | |
| AK146082.1 | - | 3 - 4 | 356 | 63 | ||
| AK171245.1 | - | 3 - 4 | 358 | 63 | ||
| BC058780.1 | - | 3 - 4 | 344 | 6312202 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfand2a
Genomic Map