Assay Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159908.1 | NP_001153380.1 | 6 - 7 | 760 | 66 | |
| NM_133349.3 | NP_579927.1 | 6 - 7 | 771 | 66 | ||
| GenBank mRNA | AF224494.1 | - | 6 - 7 | 657 | 66 | |
| AK146082.1 | - | 6 - 7 | 709 | 66 | ||
| AK171245.1 | - | 6 - 7 | 711 | 66 | ||
| BC058780.1 | - | 6 - 7 | 697 | 6312202 | 66 | |
| BU840945.1 | - | 2 - 3 | 134 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135896 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135897 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135894 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063396 | BC058780.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063396 | AK196645.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063396 | AF224494.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063396 | AK146082.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063396 | NM_133349.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583587 | AK087026.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786002 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 222973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447164 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103239 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150598 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171119 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063396 | BC058780.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063396 | AK196645.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063396 | AF224494.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063396 | AK146082.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063396 | NM_133349.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135905 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05765413 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150598 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171119 | - | - |
| Whole Mouse Genome G4122A | A_52_P15536 | BC058780.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P15536 | AK196645.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P15536 | AF224494.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P15536 | AK146082.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P15536 | NM_133349.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377826 | AF224494.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111804 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123558 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171119 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150598 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04349157 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04349152 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04349158 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04349156 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04349151 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04349155 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135897 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04433427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04653176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04349154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04349157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04215558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04759735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04433426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04534557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04944277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04944276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04835467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04349160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04191583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04944271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04215553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04215563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04349151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04215556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04944275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04463169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04536769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04580474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135881 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535102 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449702_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415940_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415941_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449703_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535095 | - | - |
| Mouse Genome 430 2.0 Array | 1449703_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415940_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449703_at | - | - |
| Mouse Genome 430 2.0 Array | 1415941_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1449702_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415940_at | - | - |
| Mouse Genome 430 2.0 Array | 1449702_at | - | - |
| Mouse Genome 430 2.0 Array | 1415941_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100494_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100494_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500031974.mm.1 | TC0500003495.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031974.mm.1 | NM_001159908.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031974.mm.1 | NM_001159908.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031974.mm.1 | NM_133349.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031974.mm.1 | NM_133349.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031975.mm.1 | TC0500003495.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031975.mm.1 | NM_001159908.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031975.mm.1 | NM_001159908.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031975.mm.1 | NM_133349.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031975.mm.1 | NM_133349.3 | Refseq |
| Mouse Expression Set 430 | 1415941_s_at | - | - |
| Mouse Expression Set 430 | 1415941_PM_s_at | - | - |
| Mouse Expression Set 430 | 1415940_at | - | - |
| Mouse Expression Set 430 | 1415940_PM_at | - | - |
| Murine Genome U74v2 Set | 161376_PM_f_at | - | - |
| Murine Genome U74v2 Set | 160713_at | - | - |
| Murine Genome U74v2 Set | 161376_f_at | - | - |
| Murine Genome U74v2 Set | 160713_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415941_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415940_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415941_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415940_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415940_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415941_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449703_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415940_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449703_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449702_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415941_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449702_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 672753 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2940112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002940112 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230489 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230489 | - | - |
| mouse-6 v1.1 (Illumina) | scl25844.10_237-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2940112 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25844.10_237-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1230489 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230489 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25844.10_237-S | - | - |

Zfand2a
Genomic Map